missing translation for 'onlineSavingsMsg'
Learn More

Brady™ EMEA SFID Suite for Brady Workstation

Product Code. 30281105
Click to view available options
:
EMEA SFID Suite
This item is not returnable. View return policy

Product Code. 30281105

Brand: Brady™ 149226

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Keep your facility safe and compliant with the EMEA SFID Suite for Brady Workstation. This group of apps makes it easy to create custom signs, labels, and pipemarkers to add visual safety guidance to your facility.

  • For Use With (Equipment): BBP11, BBP12, BBP30, BBP31, BBP33, BBP35, BBP37, BBP72, BBP81, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, J2000, J4000, J5000, i3300, i5100, i7100, i7500, M611, PR Plus, S3000, S3100, GlobalMark 2 Multicolor, Inkjet Printers, IP Series, Laser Printers, THT 76 mm Core Printers, Wraptor A6500, Wraptor, M410, M510

Specifications

Product Type Software
For Use With (Equipment) Brady Label Printers
Format Email with Activation Code
Languages Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English (US), Estonian, Finnish, French, German, Hungarian, Italian, Japanese, Korean, Norwegian (Bokmål), Polish, Portuguese, Romanian, Russian, Slovak, Slovenian, Spanish (Mexico), Spanish, Swedish, Turkish, Vietnamese
Software Type Sign and Label Design Software
Alternate Part Number BWS-SFIDS-EMEA-EM
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.