missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Emetine dihydrochloride

Product Code. 18729416
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18729416 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18729416 Supplier R&D Systems™ Supplier No. 7342/50
  • ONLINE EXCLUSIVE PRICE
    166.77 €
    149.10 € / 50mg
    Save 17.67 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. RNA polymerase inhibitor; inhibits replication of a broad range of RNA and DNA viruses, including SARS-CoV-2 and Zika

Emetine is a RNA polymerase inhibitor. It inhibits SARS-CoV-2 replication (EC50 = 460 nM) and in combination with Remdesivir (Cat. No.7226) synergistically inhibits the viral yield of SARS-CoV-2 by 64.9%. Emetine attenuates hCoV-OC43, hCoV-NL43, SARS-CoV, MERS-CoV, and MHV-A59 in vitro with EC50 values in the low micromolar range. It also inhibits replication of a broad range of RNA and DNA viruses, including influenza, ebolavirus and HIV-1. Emetine potently inhibits ZIKV replication in vitro (IC50 = 52.9 nm) and suppresses ZIKV viral load in vivo. Anti-inflammatory, antiviral, antimalarial, antibacterial and anti-amebic.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2 S,3 R,11 bS)-3-Ethyl-1,3,4,6,7,11 b-hexahydro-9,10-dimethoxy-2-[[(1 R)-1,2,3,4-tetrahydro-6,7-dimethoxy-1-isoquinolinyl]methyl]-2 H-benzo[a]quinolizine dihydrochloride
CAS 316-42-7
Quantity 50 mg
Product Line Tocris
Target RNA/DNA Polymerase Inhibitors
Molecular Formula C29H40N2O40.2 HCl
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.