missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ EMM, Powdered Media

Product Code. 11317859 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
227 g
454 g
1000 g
Unit Size:
1kg
227g
454g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317859 227 g 227g
11327859 454 g 454g
11337859 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11317859 Supplier MP Biomedicals™ Supplier No. 114110012
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Defined medium for the growth of S. pombe

Formulation

  • Contents/liter: 3 g phthalic Acid K+, 2.2 g Na2HPO4, 5 g NH4Cl, 20 g dextrose
  • Salts: 1.05 g MgCl2·6H2O, 14.7 μg CaCl2·2H2O, 1 g KCl, 0.04 g Na2SO4
  • Vitamins: 1 mg pantothenic acid, 10 mg nicotinic acid, 10 mg myo-inositol, 10 μg biotin
  • Minerals: 0.5 mg boric acid, 0.4 mg MnSO4, 0.4 mg ZnSO4·7H2O, 0.2 mg FeCl2·6H2O, 40 μg molybdic acid, 0.1 mg KI, 40 μg CuSO4·5H20, 1 mg citric acid

 

Applications:

Yeast cell culture, Life sciences

Specifications

Certifications/Compliance Certified Thiamine (Vitamin B1) free
Content And Storage 15° to 30°C
Form Powder
Grade Molecular biology
Product Type EMM
Formulation phthalic Acid K+ 3 g/L, Na2HPO4 2.2 g/L, NH4Cl 5 g/L, Dextrose 20 g/L, MgCl2.6H2O 1.05 g/L, CaCl2.2H2O 14.7 μg/L, KCl 1 g/L, Na2SO4 0.04 g/L, Pantothenic Acid 1 mg/L, Nicotinic Acid 10 mg/L, myo-inositol 10 mg/L, Biotin 10 μg/L, Boric Acid 0.5 mg/L, MnSO4 0.4 mg/L, ZnSO4.7H2O 0.4 mg/L, FeCl2.6H2O 0.2 mg/L, Molybdic Acid 40 μg/L, KI 0.1 mg/L, CuSO4.5H2O 40 μg/L, Citric Acid 1 mg/L
For Use With (Application) For growth of fission Yeast S. Pombe
Quantity 227 g
compliance-icons
Product Identifier
  • EMM Broth (Powder)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ammonium chloride, sodium phosphate dibasic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.