missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Emodin, 95%, tech.

Product Code. 10386980 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10386980 100 mg Glass bottle 100mg
10704421 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10386980 Supplier Thermo Scientific Chemicals Supplier No. 117751000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 518-82-1 | C15H10O5 | 270.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 518-82-1
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.23
MDL Number MFCD00001207
InChI Key RHMXXJGYXNZAPX-UHFFFAOYSA-N
Synonym emodin, schuttgelb, emodol, frangula emodin, rheum emodin, frangulic acid, 3-methyl-1,6,8-trihydroxyanthraquinone, archin, persian berry lake, 1,3,8-trihydroxy-6-methyl-9,10-anthraquinone
PubChem CID 3220
ChEBI CHEBI:42223
IUPAC Name 1,3,8-trihydroxy-6-methylanthracene-9,10-dione
SMILES CC1=CC(=C2C(=C1)C(=O)C3=CC(=CC(=C3C2=O)O)O)O

Specifications

Melting Point 253.0°C
Color Orange-Red
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Beilstein 08, 520
Merck Index 15, 3618
Formula Weight 270.23
Percent Purity 95%
Grade Technical
Physical Form Powder
Chemical Name or Material Emodin, tech., 95%
Show More Show Less
compliance-icons
Product Identifier
  • Emodin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.