missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Empty FastPrep™ 2 mL Lysing Matrix Tubes

Product Code. 11452350 Shop All MP Biomedicals Products
Click to view available options
Volume (Metric):
2 mL
Unit Size:
Pack of 5000
This item is not returnable. View return policy

Product Code. 11452350

Brand: MP Biomedicals™ 115076600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Available empty for those who wish to fill their own tubes. Empty FastPrep™ 2 mL Lysing Matrix Tubes (non-skirted, no caps) are 2 mL tubes that work with FastPrep or other bead-beater instruments to lsye, grind or homogenize samples.

  • Used in FastPrep systems.
  • Add sample to empty tube, add appropriate Lysing Matrix material and lysing buffer, and apply cap.
  • Empty FastPrep 2 mL tubes are screw cap tubes and can be used with any color cap.
  • They are designed for grinding and lysing samples with FastPrep instruments.
  • Non-skirted
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Volume (Metric) 2 mL
Storage Requirements Room Temperature (15°C to 30°C)
Quantity 5000 x 2 mL
Product Type Empty Lysing Matrix Tubes

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.