missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Empty Spin Columns; 50 columns

Product Code. 17577466 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
50 Columns
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17577466 50 Columns Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17577466 Supplier IBA Lifesciences Supplier No. 25150050
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Empty Spin columns for rapid protein purification with Strep-Tactin™XT 4Flow™ high capacity.

  • Empty Spin Columns are ideal for rapid protein purification from small sample volumes.
  • They are provided with inserted frits (mesh size 10 μm), screw caps, and snap-off plugs and compatible with conventional 1.5 mL or 2.0 mL reaction tubes.
  • The frit in the lower part of the column ensure that the resin remain in the spin column while sample or buffer flow through rapidly during the centrifugation process.
  • The maximum filling volume is 700 μL.
  • The empty spin columns are offered without resin and purification buffer to facilitate a customer-specific application and experimental design, e.g., if specific buffer conditions are required.
  • For purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins, the combination with Strep-Tactin™XT 4Flow™ high capacity resin is recommended.

Specifications

Workflow Step Protein purification
Quantity 50 Columns
Sample Volume max. 700 μL
Format Spin Column
Column Type Affinity, Agarose Resin

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.