missing translation for 'onlineSavingsMsg'
Learn More

Emtricitabine S-Oxide (Mixture of Diastereomers), TRC

Product Code. 30136619
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136619 100 mg 100mg
17992385 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136619 Supplier Toronto Research Chemicals Supplier No. TRCE525030100MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152128-77-3
Molecular Formula C8 H10 F N3 O4 S
Synonym 2(1H)-Pyrimidinone, 4-amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-3-oxido-1,3-oxathiolan-5-yl]-∣2(1H)-Pyrimidinone, 4-amino-5-fluoro-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-, S-oxide, [2R-(2α,5α)]-∣2(1H)-Pyrimidinone, 4-amino-5-fluoro-1-[2-(hydroxymethyl)-3-oxido-1,3-oxathiolan-5-yl]-, [2R-(2α,5α)]- (9CI)∣1,3-Oxathiolane, 2(1H)-pyrimidinone deriv.∣4-Amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-3-oxido-1,3-oxathiolan-5-yl]-2(1H)-pyrimidinone

Specifications

Format Neat
Molecular Weight (g/mol) 263.25
Shipping Condition Room Temperature
Purity >95% (HPLC)
Quantity 100 mg
Chemical Name or Material Emtricitabine S-Oxide (Mixture of Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.