Learn More
SPEX Certiprep™ Extractor for Small Vials
Product Code. 15360492 Shop All SPEX Certiprep Products
Change view
Click to view available options
Unit Size:
Each
Product Code. unitSize
15360492 Each
1 options
Product Code. 15360492
Brand: SPEX Certiprep™ 6756
This item requires a price update prior to being ordered.
Request a Price for this item here.
Use this to easily removes the end plug from frozen vials.
  • This is an end-plug extractor bench-top accessory tool for the SPEX SamplePrep freezer mill small grinding vials (6751, 6761, 6771 and 6781S). 
  • Simply place a vial into the extractor cradle and press down the extractor handle to remove the end plug of the vial.
  • Sample can be retrieved from the vial while it's still cold.
  • The extractor features suction cups that secure the extractor to the benchtop allowing for one-handed operation.
Specifications
Container Size 1