missing translation for 'onlineSavingsMsg'
Learn More

endo-(±)-Norborneol, 92%, Thermo Scientific™

Product Code. 10236000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
Product Code. Quantity Packaging unitSize
10236000 250 mg Glass bottle 250mg
1 options
This item is not returnable. View return policy

Product Code. 10236000

Brand: Thermo Scientific Acros 181462500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 497-36-9
Melting Point 148.0°C to 154.0°C
Assay Percent Range (exo-Norborneol) Remainder (GC)
Packaging Glass bottle
Molecular Formula C7H12O
MDL Number MFCD00167747,MFCD00063677
Quantity 250 mg
InChI Key ZQTYQMYDIHMKQB-UHFFFAOYNA-N
SMILES OC1CC2CCC1C2
IUPAC Name bicyclo[2.2.1]heptan-2-ol
Molecular Weight (g/mol) 112.17
Formula Weight 112.17
Percent Purity 92%
Chemical Name or Material endo-(±)-Norborneol, 92%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.