missing translation for 'onlineSavingsMsg'
Learn More

Endothall monohydrate, TRC

Product Code. 30379209
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379209 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379209 Supplier Toronto Research Chemicals Supplier No. TRCE55543310MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Endothall monohydrate
CAS 62059-43-2
Purity Grade Notes HPLC
Molecular Formula C8H10O5 . H2O
Quantity 10 mg
Synonym Endothall Monohydrate, 7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic Acid Hydrate
InChI Formula InChI=1S/C8H10O5.H2O/c9-7(10)5-3-1-2-4(13-3)6(5)8(11)12;/h3-6H,1-2H2,(H,9,10)(H,11,12);1H2
SMILES O.OC(=O)C1C2CCC(O2)C1C(=O)O
Molecular Weight (g/mol) 204.18
Formula Weight 204.0634
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.