missing translation for 'onlineSavingsMsg'
Learn More

R&DSystems™ Endothelial Cell Growth Supplement without VEGF (50X)

Product Code. 18441950
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18441950 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18441950 Supplier R&DSystems™ Supplier No. 390634
  • ONLINE EXCLUSIVE PRICE
    105.38 €
    92.40 € / 5mL
    Save 12.98 €
    12% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Media Supplement for the Low-serum, VEGF-free Expansion of Primary Endothelial Cells

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Line R&D Systems
Quantity 5 mL
Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.