missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Endotoxin Binding Reagent

Product Code. 14889720 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
60 mL
Unit Size:
60mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
14889720 60 mL 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 14889720 Supplier Thermo Scientific™ Supplier No. R1601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific GeneJET Endotoxin Binding Reagent is a component of the GeneJET Plasmid Maxiprep Kit and may be purchased separately. Provided in 60 mL aliquots.

Thermo Scientific GeneJET Endotoxin Binding Reagent is a component of the GeneJET Plasmid Maxiprep Kit (Cat. No. K0491/K0492) and may be purchased separately. Provided in 60 mL aliquots.

Thermo Scientific GeneJET Endotoxin Binding Reagent is a component of the GeneJET Plasmid Maxiprep Kit (Cat. No. K0491/K0492) and may be purchased separately. Provided in 60 mL aliquots.
TRUSTED_SUSTAINABILITY

Specifications

Product Line GeneJET
Product Type Endotoxin Binding Reagent
Quantity 60 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.