missing translation for 'onlineSavingsMsg'
Learn More

Endotoxin-free Water, Waternation

Product Code. 31921344 Shop All Waternation Medical Division Products
Change view
Click to view available options
Quantity:
1 L
5 L
6 L
20 L
Unit Size:
1 L
20 L
5 L
6 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31921344 1 L 1 L
31921349 5 L 5 L
31921346 6 L 6 L
31921343 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31921344 Supplier Waternation Supplier No. WNEN10010
  • ONLINE EXCLUSIVE PRICE
    78.25 €
    55.09 € / 1 L
    Save 23.16 €
    29% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Endotoxin-free water is specifically designed for use in applications that require extremely low endotoxin levels, including cell culture, protein work, molecular biology, and pharmaceutical or medical device testing.

It is important to use high-resistivity purified water (18.2 MΩ) with very low organic content (TOC ≤15 ppb) and free of endotoxins. Endotoxins can significantly alter cell growth and morphology - for example, in cardiomyocytes and may also subtly affect cell function. Working with cultures unknowingly contaminated with endotoxins can lead to inaccurate results and wasted time, money, and effort. Using Endotoxin Free water can effectively prevent this risk.

  • Ultra Pure Water is sterile filtered through a 0.05-micron filter and is considered endotoxin-free (<0.005 EU/mL)
  • Sterility Testing: Negative for Growth
  • pH: 6-7
  • The product is available in a range of sizes and packaging options.

Specifications

Molecular Weight (g/mol) 18.015
Percent Purity 99.99%
Grade Cell Culture/Molecular Biology, EP/USP, Biotech, Ultrapure Bioreagent, Certified
Quantity 1 L
Chemical Name or Material Pure water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.