missing translation for 'onlineSavingsMsg'
Learn More

Endotoxin Testing

Product Code. 10594034
Change view
Click to view available options
Content And Storage:
Store at room temperature.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Unit Size
10594034 Store at room temperature. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10594034 Supplier Invitrogen Supplier No. 100002314
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.