missing translation for 'onlineSavingsMsg'
Learn More

Enmetazobactam, MedChemExpress

Product Code. 16476517
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476517 5 mg 5mg
16474846 100 mg 100mg
16423798 10 mg 10mg
16420177 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476517 Supplier Medchem Express Supplier No. HY103095
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Enmetazobactam (AAI101) is an extended-spectrum β-lactamase inhibitor, against many resistant Gram-negative pathogens.

Enmetazobactam (AAI101) is an extended-spectrum β-lactamase inhibitor, against many resistant Gram-negative pathogens.

Tekniske data

Chemical Name or Material Enmetazobactam
CAS 1001404-83-6
Color White
Purity Grade Notes Research
Molecular Formula C11H14N4O5S
Quantity 5 mg
Synonym AAI101
Solubility Information DMSO : ≥ 113.3 mg/mL (360.46 mM)
SMILES C[N+]1=NN(C[C@@](S([C@]2([H])C3)(=O)=O)(C)[C@H](C([O-])=O)N2C3=O)C=C1
Molecular Weight (g/mol) 314.32
Formula Weight 314.32
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.