missing translation for 'onlineSavingsMsg'
Learn More

Enoxacin, TRC

Product Code. 30134407
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134407 5 g 5g
30133433 1 g 1g
30008728 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134407 Supplier Toronto Research Chemicals Supplier No. TRCE5576005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74011-58-8
Molecular Formula C15 H17 F N4 O3
Molecular Weight (g/mol) 320.32
Synonym 1,8-Naphthyridine-3-carboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)- (9CI, ACI), 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-naphthyridine-3-carboxylic acid (ACI), 1-Ethyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid, 1-Ethyl-6-fluoro-4-oxo-7-piperazin-4-ium-1-yl-1,8-naphthyridine-3-carboxylate, AT 2266, CI 919, Enofloxacin, Enofloxacine, Enoksetin, Enoxacine, ENOXOR, Flumark, MeSH ID: D015365, NSC 629661, PD 107779, Penetrex, Penetrex (pharmaceutical)
IUPAC Name 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid
SMILES CCN1C=C(C(=O)O)C(=O)c2cc(F)c(nc12)N3CCNCC3

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H17FN4O3/c1-2-19-8-10(15(22)23)12(21)9-7-11(16)14(18-13(9)19)20-5-3-17-4-6-20/h7-8,17H,2-6H2,1H3,(H,22,23)
Formula Weight 320.1285
Percent Purity >95%
Chemical Name or Material Enoxacin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.