missing translation for 'onlineSavingsMsg'
Learn More

Ensulizole, TRC

Product Code. 30381537
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381537 10 g -
30382879 50 g 50g
30374928 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381537 Supplier Toronto Research Chemicals Supplier No. TRCE55790010G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27503-81-7
Molecular Formula C13 H10 N2 O3 S
Molecular Weight (g/mol) 274.3
Synonym 2-Phenyl-1H-Benzimidazole-5-sulfonic acid, 2-Phenyl-5-Benzimidazolesulfonic acid, 2-Phenyl-5-sulfobenzimidazole, 2-Phenylbenzimidazole-5-sulfonic acid, Ensulizole, Eusolex 232, Neo Heliopan Hydro, Novantisol, Parsol HS, Phenylbenzimidazolesulfonic acid
IUPAC Name 2-phenyl-1H-benzimidazole-5-sulfonic acid
SMILES OS(=O)(=O)c1ccc2[nH]c(nc2c1)c3ccccc3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H10N2O3S/c16-19(17,18)10-6-7-11-12(8-10)15-13(14-11)9-4-2-1-3-5-9/h1-8H,(H,14,15)(H,16,17,18)
Formula Weight 274.0412
Chemical Name or Material Ensulizole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.