missing translation for 'onlineSavingsMsg'
Learn More

ent-Emtricitabine, TRC

Product Code. 30383413
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383413 1 mg
30376428 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383413 Supplier Toronto Research Chemicals Supplier No. TRCE5250051MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137530-41-7
Molecular Formula C8 H10 F N3 O3 S
Molecular Weight (g/mol) 247.25
Synonym 2(1H)-Pyrimidinone, 4-amino-5-fluoro-1-[(2S,5R)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-, 2(1H)-Pyrimidinone, 4-amino-5-fluoro-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-, (2S-cis)-, 1,3-Oxathiolane, 2(1H)-pyrimidinone deriv., 4-Amino-5-fluoro-1-[(2S,5R)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-2(1H)-pyrimidinone, (+)-2'-Deoxy-3'-thia-5-fluorocytidine, (+)-FTC, (2'S,5'R)-Emtricitabine, 4-Amino-5-fluoro-1-[(2S,5R)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2(1H)-one
IUPAC Name 4-amino-5-fluoro-1-[(2S,5R)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1F)[C@H]2CS[C@@H](CO)O2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C8H10FN3O3S/c9-4-1-12(8(14)11-7(4)10)5-3-16-6(2-13)15-5/h1,5-6,13H,2-3H2,(H2,10,11,14)/t5-,6+/m1/s1
Formula Weight 247.0427
Chemical Name or Material ent-Emtricitabine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.