missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Entecavir
Description
Entecavir is a potent and selective hepatitis B virus reverse transcriptase inhibitor (EC50 = 3 nM); guanine analog. Inhibits hepatitis B virus replication in HepG2.2.15 cells. Displays selectively over HCMV, HSV-1, VZV, HIV and influenza (EC50 values are 15, 32, 30-60, >10 and >80 μM respectively). Orally bioavailable.
Specifications
Specifications
| CAS | 142217-69-4 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Molecular Formula | C12H15N5O3 |
| Formula Weight | 277.28 |
| Product Type | Inhibitor |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
¿Detecta una oportunidad de mejora?