missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Enterobacteria Enrichment Broth (Dehydrated)
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
Selectively enrich bile tolerant Gram-negative bacteria during microbiological examination of pharmaceutical products with Thermo Scientific™ Oxoid™ Enterobacteria Enrichment Broth-Mossel (EP/USP/JP/BP).
Enterobacteria Enrichment Broth-Mossel (EP/USP/JP/BP) is recommended as an enrichment medium for bile-tolerant Gram-negative bacteria in the microbiological examination of pharmaceutical products. It can be used as test for absence or as a quantitative test1.
Enterobacteria Enrichment Broth-Mossel (EP/USP/JP/BP) should be used as an enrichment broth in conjunction with Violet Red Bile Glucose Agar (R455302).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
- Adheres to EP/USP/JP/BP specifications.
Enterobacteria Enrichment Broth-Mossel (EP/USP/JP/BP) should be used as an enrichment broth in conjunction with Violet Red Bile Glucose Agar (R455302).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
Specifications
| Content And Storage | 10°C to 30°C |
| Description | Enterobacteria Enrichment Broth (dehydrated) |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction