Learn More
Memmert™ Entry Port 14 mm for IF750plus
Product Code. 17527215 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF750plus Incubator
Unit Size:
Each
Product Code. 17527215
Brand: Memmert™ IF750plusD6
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Incubators.
  • Entry port with 14 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) IF750plus Incubator
Type Entry Port
Diameter (Metric) 14 mm