Description
- Entry port with 23 mm diameter
- Flap closure
- Custom position TBC in right side
Specifications
Specifications
| For Use With (Equipment) | UF55 Oven |
| Type | Entry Port |
| Diameter (Metric) | 23 mm |