Learn More
Memmert™ Entry Port 57 mm for IF450plus
Product Code. 17557045 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
IF450plus Incubator
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17557045 IF450plus Incubator Each
1 options
Product Code. 17557045 Supplier Memmert™ Supplier No. IF450plusF8
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Incubators.
  • Entry port with 57 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) IF450plus Incubator
Type Entry Port
Diameter (Metric) 57 mm