missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Entry Port Right Centre/Centre

Product Code. 17110275 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17110275

Brand: Memmert™ IPP750ecoF2

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For introducing connections at the side and can be closed by flap

  • Standard position right centre/centre
  • Not for model sizes 110 and 260 with light module
  • Not for model size 110 with light module

Specifications

Color Clear
For Use With (Equipment) IPP750eco
Diameter (Metric) 23 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.