missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Caspase-3 Activity Assay Kits

Product Code. 10174472
Change view
Click to view available options
Label or Dye:
AMC (7-amino-4-methylcoumarin)
Rhodamine 110
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Label or Dye Unit Size
10174472 AMC (7-amino-4-methylcoumarin) Each
10573473 Rhodamine 110 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10174472 Supplier Invitrogen™ Supplier No. E13183
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Detect apoptosis in cell extracts and purified enzyme preparations with EnzChek Caspase-3 Activity Assay Kits with Z-DEVD-AMC substrate or Z-DEVD-R110 substrates.

Enable simple and reliable detection of caspase-3-activity-mediated apoptosis with the EnzChek Caspase-3 Activity Assay Kit, which takes advantage of specially designed substrates that fluoresce upon cleavage by the caspase-3 enzyme. The EnzChek Caspase-3 Activity Assay Kit is available with either the Z-DEVD-AMC or Z-DEVD-R110 substrate, which yields a bright fluorescent product upon proteolytic cleavage by caspase 3/7.

The EnzChek Caspase-3 Activity Assay Kit enables detection of apoptosis by providing a simple and reliable method for assaying caspase-3/7 activity. The kit can be used to continuously measure the activity of caspase-3/7 in cell extracts and purified enzyme preparations using a fluorometer or fluorescence microplate reader.

Key features of the EnzChek Caspase-3 Activity Assay Kit includes:

  • Fluorescent assay using standard fluorescein (FITC) excitation/emission settings
  • Format allows for continuous measurement of caspase-3/7 activity in cell extracts
  • Fluorescent and enzymatic controls included

The aminomethylcoumarin (AMC)-derived substrate (Z-aspartic acid-glutamic acid-valine-aspartic acid (DEVD)-AMC) that is included in the EnzChek Caspase-3 Activity Assay Kit is weakly fluorescent in the UV range (excitation/emission maxima ∼330/390 nm), but yields a bright, blue-fluorescent product (excitation/emission maxima ∼342/441 nm) upon proteolytic cleavage. The kit can be used to continuously measure the activity of caspase-3 and closely related proteases in cell extracts and purified enzyme preparations using a fluorometer or fluorescence microplate reader.

The rhodamine 110-derived substrate (Z-DEVD-R110) used in the EnzChek Caspase-3 Activity Assay Kit is a non-fluorescent bisamide compound that, upon enzymatic cleavage, is converted in a two-step process to the fluorescent monoamide and then to the even more fluorescent R110 product. Both of these hydrolysis products exhibit spectral properties similar to those of fluorescein, with peak excitation and emission wavelengths of 496 nm and 520 nm, respectively.

In addition to the substrates, the EnzChek Caspase Activity Assay Kit contains the reversible aldehyde inhibitor Ac-DEVD-CHO, as well as an AMC or R110 reference standard. The Ac-DEVD-CHO inhibitor confirms that the fluorescence signals in both induced and control cell populations are due to the activity of the caspase-3/7 protease. The reference standard allows for quantification of the amount of AMC or R110 released in the reaction.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Description EnzChek Caspase-3 Assay Kit #1, Z-DEVD-AMC substrate
Quantity 500 Assays
Format Cuvettes, 96-well Plate
Product Type Caspase Assay Kit
Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Excitation/Emission 342/441 (cleaved substrate)
No. of Reactions 500 Assays (reaction volume 100μL per assay)
Shipping Condition Room Temperature
Product Line EnzChek
Detection Method Fluorescence
For Use With (Equipment) Fluorometer, Microplate Reader
Label or Dye AMC (7-amino-4-methylcoumarin)
Label Type Other Label(s) or Dye(s)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.