missing translation for 'onlineSavingsMsg'
Learn More

EOAI3402143, MedChemExpress

Product Code. 16495326
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495326 100 mg 100mg
16487167 5 mg 5mg
16480757 50 mg 50mg
16463086 25 mg 25mg
16444448 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495326 Supplier Medchem Express Supplier No. HY111408

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EOAI3402143 is a deubiquitinase (DUB) inhibitor, which inhibits dose-dependently inhibits Usp9x/Usp24 and Usp5.

EOAI3402143 is a deubiquitinase (DUB) inhibitor, which inhibits dose-dependently inhibits Usp9x/Usp24 and Usp5.

Specifications

Chemical Name or Material EOAI3402143
CAS 1699750-95-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H28Cl2N4O3
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (99.32 mM; Need ultrasonic)
SMILES ClC1=C(/C=C(C(NC(CCC)C2=CC=C(OCCN3CCOCC3)C=C2)=O)\C#N)N=C(Cl)C=C1.[E]
Molecular Weight (g/mol) 503.42
Formula Weight 503.42
Percent Purity 99.52%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.