missing translation for 'onlineSavingsMsg'
Learn More

Eosin B, pure, certified

Product Code. 10404315 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
Product Code. Quantity Packaging unitSize
10404315 100 g Glass bottle 100g
10404891 25 g Glass bottle 25g
2 options
This item is not returnable. View return policy

Product Code. 10404315

Brand: Thermo Scientific Acros 409421000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 548-24-3 | C20H6Br2N2Na2O9

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-24-3
Molecular Formula C20H6Br2N2Na2O9
MDL Number MFCD00005041
Synonym 4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, Eosin Bluish

Specifications

Absorption Ratio 0.96 to 1.22
Dye Content 85% min.
Beilstein 19, 233
Merck Index 15, 3657
Infrared Spectrum Authentic
Lambda Max 511 to 520nm (in water + 1% Na2CO3)
Formula Weight 624.06
Grade Pure
Packaging Glass bottle
Color Brown to Purple/Red
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material Eosin B, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Eosin B
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Eosin B

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.