missing translation for 'onlineSavingsMsg'
Learn More

Eosin Y Disodium Salt, 10% Solution, MP Biomedicals™

Product Code. 11482511 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
10 L
Unit Size:
1 L
10 L
100mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482511 10 L 10 L
11452511 100 mL 100mL
11462511 500 mL 500mL
11472511 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11482511 Supplier MP Biomedicals Supplier No. 0219570010
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reduces the affinity of Mg2+-ATPase for ATP and increases the enzyme affinity for Mg2+. Eosin Y Disodium Salt, 10% Solution, MP Biomedicals is a red xanthene dye. It is a photosensitizer that works as a molecular photoelectrode and it enhances fluorescent

At concentrations higher than 10-50μM (near 100μM) the affinity for Mg2+-ATPase for the ion-activator is reduced. This compound works as a molecular photoelectrode via catalyzation of the visible-light-driven electron-transfer reaction. This agent also enhances fluorescent quenching abilities by forming a complex with polyvinulpyrrolidone. It is used as a counterstain for hematoxylin.

Eosin Y disodium salt is used in combination with Azure II: to show morphological details of marrow cells; buffered Azure A/Eosin Y method for demostrating cell granules, microorganisms and nuclei; for showing insect musculature; for demonstrating cells infected with mosquito-iridescent virus; use in combination with Alum Hematoxylin/Azure II to show mast cells, early necrobiotic changes in cells and protozoans, rickettsias and bacteria in tissues; to demonstrate volutin granules in any microorganism; Bond's modification of Mann's Eosin-Methyl Blue for Negri bodies in smears; Harris stain for Negri bodies in smears; Mann's method for staining Negri bodies in sections; Gridley's stain for Endamoeba in tissue sections; a modified methenamine-silver nitrate method for the demonstration of Endamoeba in sections.

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H12Br4Na2O8
Molecular Weight (g/mol) 745.904
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Synonym 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Acid red 87, Eosin yellowish solution, Eosin Y solution, Eosin Y
PubChem CID 91886399
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]

Specifications

Formula Weight 691.86
Quantity 10 L
Physical Form Solution
compliance-icons
Product Identifier
  • Eosin Y Disodium Salt, 10% Solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: Eosin Y Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.