missing translation for 'onlineSavingsMsg'
Learn More

Eosin Y Disodium Salt, MP Biomedicals

Product Code. 11462742 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462742 25 g 25g
11472742 100 g 100g
11482742 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462742 Supplier MP Biomedicals Supplier No. 0215247125
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H12Br4Na2O8
Molecular Weight (g/mol) 745.904
MDL Number MFCD00005040
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Synonym 2',4',5',7'-Tetrabromofluorescein disodium salt, 2', 4', 5', Acid Red 87
PubChem CID 91886399
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]

Specifications

Formula Weight 691.86
Color Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Eosin Y Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.