missing translation for 'onlineSavingsMsg'
Learn More

Epiandrosterone, 99%, Thermo Scientific Chemicals

Product Code. 10431641 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10431641

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Epiandrosterone, 99%, CAS# 481-29-8, is a steroid hormone with weak androgenic activity. It is a metabolite and a precursor of testosterone and estradiol with hypolipidemic and anabolic activities.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Epiandrosterone is a steroid hormone, derived from 5 α-androstane

• It is a dehydroepiandrosterone metabolite and an androgen

• It is a precursor of testosterone and estradiol with hypolipidemic and anabolic activities

Applications

• Epiandrosterone presents some neurological activity since it can bind to the ɣ-aminobutyric acid (GABA)/benzodiazepine-receptor complex (GABA-RC), acting as a negative non-competitive modulator of GABA-RC, and signaling through the N-methyl-D-aspartate receptor

• It can also inhibit the pentose phosphate pathway, dilating blood vessels pre-contracted by partial depolarization

• Epiandrosterone can inhibit the thromboxane A2 synthesis in activated platelets, decrease plasma plasminogen activator inhibitor type 1, reduce tissue plasminogen activator antigen, improve insulin-like growth factor 1 serum levels, and increase the synthesis of cyclic guanosine monophosphate and nitric oxide

• In vitro assays demonstrate that this compound inhibits Junin virus replication vitro anti-adenovirus (AdV) activity

• Epiandrosterone affects glucose oxidation and interleukin-1 beta in in vitro pancreatic islets

TRUSTED_SUSTAINABILITY

Specifications

CAS 481-29-8
Melting Point 171°C to 177°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C19H30O2
MDL Number MFCD00064134
Quantity 5 g
Specific Rotation Condition +91° (20°C c=1,CH3OH)
Specific Rotation +88° to +94° (20°C, 589nm) (c=1, CH3OH)
Synonym epiandrosterone, trans-androsterone, isoandrosterone, 3beta-hydroxy-5alpha-androstan-17-one, 3beta-hydroxyetioallocholan-17-one, epi-andosterone, 5alpha-androstan-3beta-ol-17-one, iso-androsterone, d-epiandrosterone, 3-epiandrosterone
InChI Key QGXBDMJGAMFCBF-LUJOEAJASA-N
SMILES CC12CCC(CC1CCC3C2CCC4(C3CCC4=O)C)O
IUPAC Name (3S,5S,8R,9S,10S,13S,14S)-3-hydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-one
Molecular Weight (g/mol) 290.447
PubChem CID 441302
ChEBI CHEBI:541975
Formula Weight 290.44
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material Epiandrosterone, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.