missing translation for 'onlineSavingsMsg'
Learn More

(±)-Epichlorohydrin, 99%

Product Code. 11412498
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412498 100 g 100g
11422498 500 g 500g
11432498 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11412498 Supplier Thermo Scientific Alfa Aesar Supplier No. A15823.22
 Restricted Item
In Stock
Add to basket
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Normativo: Questo prodotto è un intermedio isolato trasportato in conformità con il regolamento (CE) 1907/2006 (REACH). Tutti i clienti devono compilare una dichiarazione prima della consegna.

CAS: 106-89-8 | C3H5ClO | 92.522 g/mol

Epichlorohydrin is used in the manufacture of polyamines and polyquaternary ammonium salts, as flocculents in water and wastewater treatment. It is used in the synthesis of surface active agents for washing products and toiletries. Polyamine-epichlorohydrin resin is used in paper industry to improve paper wet-strength. It acts as a chemical intermediate and widely used in the manufacture of epoxy resins, epichlorohydrin elastomers and polyamide-epichlorohydrin resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Epichlorohydrin is used in the manufacture of polyamines and polyquaternary ammonium salts, as flocculents in water and wastewater treatment. It is used in the synthesis of surface active agents for washing products and toiletries. Polyamine-epichlorohydrin resin is used in paper industry to improve paper wet-strength. It acts as a chemical intermediate and widely used in the manufacture of epoxy resins, epichlorohydrin elastomers and polyamide-epichlorohydrin resins.

Solubility
Miscible with water and organic polar solvents.

Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 106-89-8
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.522
MDL Number MFCD00005132
InChI Key BRLQWZUYTZBJKN-UHFFFAOYSA-N
Synonym epichlorohydrin, 2-chloromethyl oxirane, epichlorhydrin, 1-chloro-2,3-epoxypropane, glycidyl chloride, oxirane, chloromethyl, epichlorhydrine, 1,2-epoxy-3-chloropropane, 2,3-epoxypropyl chloride, chloropropylene oxide
PubChem CID 7835
ChEBI CHEBI:37144
IUPAC Name 2-(chloromethyl)oxirane
SMILES C1C(O1)CCl

Specifica

Melting Point -57°C
Density 1.182
Boiling Point 115°C to 117°C
Flash Point 32°C (90°F)
Assay Percent Range 99%
Odor Like Chlorine
Refractive Index 1.438
Quantity 100 g
UN Number UN2023
Beilstein 79785
Merck Index 14,3611
Solubility Information Miscible with water and organic polar solvents.
Formula Weight 92.53
Percent Purity 99%
Chemical Name or Material (+/-)-Epichlorohydrin
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons compliance-icons compliance-icons
Identificativo prodotto
  • (+/-)-Epichlorohydrin
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Indicazioni di pericolo
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
Consigli di prudenza
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informazioni supplementari
  • MIXTURE LIST-Contain: Epichlorohydrin

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.