missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (-)-Epigallocatechin gallate, 95%

Product Code. 15214248 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 15214248

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 989-51-5 | C22H18O11 | 458.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 989-51-5
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Molecular Formula C22H18O11
Quantity 10 mg
Merck Index 153573
Synonym =--epigallocatechin gallate, egcg, epigallocatechin gallate, epigallocatechin 3-gallate, tea catechin, epigallocatechin-3-gallate, =--epigallocatechin-3-o-gallate, teavigo, =--epigallocatechol gallate, epigallocatechin-3-monogallate
Solubility Information Solubility in water: partially soluble. Other solubilities: practically insoluble in acetone,dmso,methanol
InChI Key WMBWREPUVVBILR-WIYYLYMNSA-N
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O
IUPAC Name [(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
Molecular Weight (g/mol) 458.37
PubChem CID 65064
ChEBI CHEBI:4806
Formula Weight 458.37
Percent Purity 95%
Chemical Name or Material (-)-Epigallocatechin gallate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.