missing translation for 'onlineSavingsMsg'
Learn More

Epinastine Hydrochloride, TRC

Product Code. 30138113
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138113 100 mg 100mg
30135454 5 mg 5mg
30004278 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138113 Supplier Toronto Research Chemicals Supplier No. TRCE588560100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108929-04-0
Molecular Formula C16 H15 N3 . Cl H
Molecular Weight (g/mol) 285.77
Synonym (13bRS)-9,13b-Dihydro-1H-dibenzo[c,f]imidazo[1,5-a]azepin-3-amine hydrochloride, 1H-Dibenz[c,f]imidazo[1,5-a]azepin-3-amine, 9,13b-dihydro-, monohydrochloride (9CI), Alesion, Elestat, Epinastine hydrochloride, WAL 801CL
IUPAC Name (13bRS)-9,13b-Dihydro-1H-dibenzo[c,f]imidazo[1,5-a]azepin-3-amine hydrochloride
SMILES Cl.NC1=NCC2N1c3ccccc3Cc4ccccc24

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H15N3.ClH/c17-16-18-10-15-13-7-3-1-5-11(13)9-12-6-2-4-8-14(12)19(15)16;/h1-8,15H,9-10H2,(H2,17,18);1H
Formula Weight 285.1033
Chemical Name or Material Epinastine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.