missing translation for 'onlineSavingsMsg'
Learn More

Epirubicinol (Mixture of Diastereomers), TRC

Product Code. 30136750
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136750 10 mg 10mg
30004498 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136750 Supplier Toronto Research Chemicals Supplier No. TRCE58865510MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76155-56-1
Molecular Formula C27 H31 N O11
Molecular Weight (g/mol) 545.54
Synonym 5,12-Naphthacenedione, 10-[(3-amino-2,3,6-trideoxy-α-L-arabino-hexopyranosyl)oxy]-8-[(1S)-1,2-dihydroxyethyl]-7,8,9,10-tetrahydro-6,8,11-trihydroxy-1-methoxy-, (8S,10S)-, 5,12-Naphthacenedione, 10-[(3-amino-2,3,6-trideoxy-α-L-arabino-hexopyranosyl)oxy]-8-(1,2-dihydroxyethyl)-7,8,9,10-tetrahydro-6,8,11-trihydroxy-1-methoxy-, [8S-[8α,8(R*),10α]]-, (8S,10S)-10-[(3-Amino-2,3,6-trideoxy-α-L-arabino-hexopyranosyl)oxy]-8-[(1S)-1,2-dihydroxyethyl]-7,8,9,10-tetrahydro-6,8,11-trihydroxy-1-methoxy-5,12-naphthacenedione, 4'-Epiadriamycinol, Epidoxorubicinol, Epirubicinol
IUPAC Name (7S,9S)-7-[(2R,4S,5R,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-9-[(1S)-1,2-dihydroxyethyl]-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione
SMILES COc1cccc2C(=O)c3c(O)c4C[C@](O)(C[C@H](O[C@H]5C[C@H](N)[C@@H](O)[C@H](C)O5)c4c(O)c3C(=O)c12)[C@@H](O)CO

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C27H31NO11/c1-10-22(31)13(28)6-17(38-10)39-15-8-27(36,16(30)9-29)7-12-19(15)26(35)21-20(24(12)33)23(32)11-4-3-5-14(37-2)18(11)25(21)34/h3-5,10,13,15-17,22,29-31,33,35-36H,6-9,28H2,1-2H3/t10-,13-,15-,16-,17-,22-,27-/m0/s1
Formula Weight 545.1897
Chemical Name or Material Epirubicinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.