missing translation for 'onlineSavingsMsg'
Learn More

Epithiochlorohydrine, 97%

Product Code. 10033490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10033490 5 g Glass bottle 5g
10628074 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10033490 Supplier Thermo Scientific Acros Supplier No. 291390050
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3221-15-6 | C3H5ClS | 108.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3221-15-6
Molecular Formula C3H5ClS
Molecular Weight (g/mol) 108.59
MDL Number MFCD00041199
InChI Key XRWMHJJHPQTTLQ-UHFFFAOYSA-N
Synonym 2-chloromethyl thiirane, thioepichlorohydrin, epithiochlorohydrin, thioepichlorhydrin, chloropropylene sulfide, chloromethyl thiirane, 3-chloropropylene sulfide, 2-thiiranylmethyl chloride, thiirane, chloromethyl, chloropropylene sulphide
PubChem CID 18588
IUPAC Name 2-(chloromethyl)thiirane
SMILES C1C(S1)CCl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2750g/mL
Boiling Point 69°C to 70°C (67.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5240 to 1.528
Quantity 5 g
Specific Gravity 1.275
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol,chloroform,diethylether
Formula Weight 108.59
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Epithiochlorohydrine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Epithiochlorohydrine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.