missing translation for 'onlineSavingsMsg'
Learn More

Epithiochlorohydrine, 97%

Product Code. 10628074 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10628074

Brand: Thermo Scientific Acros 291390250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3221-15-6 | C3H5ClS | 108.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 3221-15-6
Molecular Formula C3H5ClS
Molecular Weight (g/mol) 108.59
MDL Number MFCD00041199
InChI Key XRWMHJJHPQTTLQ-UHFFFAOYSA-N
Synonym 2-chloromethyl thiirane, thioepichlorohydrin, epithiochlorohydrin, thioepichlorhydrin, chloropropylene sulfide, chloromethyl thiirane, 3-chloropropylene sulfide, 2-thiiranylmethyl chloride, thiirane, chloromethyl, chloropropylene sulphide
PubChem CID 18588
IUPAC Name 2-(chloromethyl)thiirane
SMILES C1C(S1)CCl

Spezifikation

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2750g/mL
Boiling Point 69°C to 70°C (67.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5240 to 1.528
Quantity 25 g
Specific Gravity 1.275
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol,chloroform,diethylether
Formula Weight 108.59
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Epithiochlorohydrine
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Epithiochlorohydrine
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Gefahrenhinweis
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H300-Fatal if swallowed.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt