missing translation for 'onlineSavingsMsg'
Learn More

Eplerenone Hydroxyacid Potassium Salt, TRC

Product Code. 30374706
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374706 5 mg
30374528 10 mg
30371519 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30374706 Supplier Toronto Research Chemicals Supplier No. TRCE5887805MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95716-98-6
Molecular Formula C24 H31 O7 . K
Molecular Weight (g/mol) 470.6
Synonym Pregn-4-ene-7,21-dicarboxylic acid, 9,11-epoxy-17-hydroxy-3-oxo-, 7-methyl ester, potassium salt (1:1), (7α,11α,17α)-, Pregn-4-ene-7,21-dicarboxylic acid, 9,11-epoxy-17-hydroxy-3-oxo-, 7-methyl ester, monopotassium salt, (7α,11α,17α)- (9CI), SC 70303, Eplerenone Hydroxyacid Potassium Salt, 9,11α-Epoxy-17-hydroxy-7α-(methoxycarbonyl)-3-oxo-17α-pregn-4-ene-21-carboxylic acid potassium salt, Pregn-4-ene-7,21-dicarboxylic acid, 9,11-epoxy-17-hydroxy-3-oxo-, 7-methyl ester, potassium salt (1:1), (7α,11α,17α)-, Pregn-4-ene-7,21-dicarboxylic acid, 9,11-epoxy-17-hydroxy-3-oxo-, 7-methyl ester, monopotassium salt, (7α,11α,17α)- (9CI), SC 70303, Eplerenone Hydroxyacid Potassium Salt, 9,11α-Epoxy-17-hydroxy-7α-(methoxycarbonyl)-3-oxo-17α-pregn-4-ene-21-carboxylic acid potassium salt
IUPAC Name potassium;3-[(1R,2S,9R,10R,11S,14R,15S,17R)-14-hydroxy-9-methoxycarbonyl-2,15-dimethyl-5-oxo-18-oxapentacyclo[8.8.0.01,17.02,7.011,15]octadec-6-en-14-yl]propanoate
SMILES [K+].COC(=O)[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@@]34O[C@@H]3C[C@@]5(C)[C@@H](CC[C@@]5(O)CCC(=O)[O-])[C@H]14

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C24H32O7.K/c1-21-7-4-14(25)10-13(21)11-15(20(28)30-3)19-16-5-8-23(29,9-6-18(26)27)22(16,2)12-17-24(19,21)31-17;/h10,15-17,19,29H,4-9,11-12H2,1-3H3,(H,26,27);/q;+1/p-1/t15-,16+,17-,19+,21+,22+,23-,24-;/m1./s1
Formula Weight 470.1707
Chemical Name or Material Eplerenone Hydroxyacid Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.