missing translation for 'onlineSavingsMsg'
Learn More

Eplerenone, MedChemExpress

Product Code. 16448438
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448438 10 mg 10mg
16434317 50 mg 50mg
16418196 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16448438 Supplier Medchem Express Supplier No. HYB0251
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Eplerenone (Epoxymexrenone) is a selective, competitive and oreally active aldosterone antagonist with an IC50 of 138 nM. Eplerenone has low affinity for progesterone, androgen, estrogen and glucocorticoid receptors. Eplerenone can be used for hypertension and heart failure after myocardial infarction reserch.

Eplerenone (Epoxymexrenone) is a selective, competitive and oreally active aldosterone antagonist with an IC50 of 138 nM. Eplerenone has low affinity for progesterone, androgen, estrogen and glucocorticoid receptors. Eplerenone can be used for hypertension and heart failure after myocardial infarction reserch.

Specifications

Chemical Name or Material Eplerenone
CAS 107724-20-9
Color White
Purity Grade Notes Research
Molecular Formula C24H30O6
Quantity 10 mg
Synonym Epoxymexrenone
Solubility Information DMSO : 25 mg/mL (60.32 mM; Need ultrasonic)
SMILES O=C(CC1)O[C@@]21CC[C@@]3([H])[C@]4([H])[C@H](C(OC)=O)CC5=CC(CC[C@]5(C)[C@@]46[C@H](O6)C[C@@]32C)=O
Molecular Weight (g/mol) 414.49
Formula Weight 414.49
Percent Purity 99.68%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.