missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Eplerenone
GREENER_CHOICE

Product Code. 18054612
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18054612 10 mg 10mg
18014302 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18054612 Supplier Tocris Bioscience™ Supplier No. 2397/10
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective mineralocorticoid receptor antagonist

Selective mineralocorticoid (aldosterone) receptor antagonist (IC50 = 360 nM). Displays > 27-fold selectivity over androgen, progesterone and estrogen receptors (IC50 > 10 μM). Orally active antihypertensive in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107724-20-9
Molecular Formula C24H30O6

Specifications

Quantity 10 mg
Formula Weight 414.5
Purity >99%
Molecular Weight (g/mol) 417.516
Chemical Name or Material Eplerenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.