missing translation for 'onlineSavingsMsg'
Learn More

Epon 828, Technical Grade, TRC

Product Code. 30384454
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384454 25 g
30373373 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384454 Supplier Toronto Research Chemicals Supplier No. TRCE58897025G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25068-38-6
Synonym Phenol, 4,4'-(1-methylethylidene)bis-, polymer with 2-(chloromethyl)oxirane (ACI), Oxirane, (chloromethyl)-, polymer with 4,4'-(1-methylethylidene)bis[phenol] (9CI), Phenol, 4,4'-(1-methylethylidene)bis-, polymer with (chloromethyl)oxirane (9CI), Phenol, 4,4'-isopropylidenedi-, polymer with 1-chloro-2,3-epoxypropane (8CI), Propane, 1-chloro-2,3-epoxy-, polymer with 4,4'-isopropylidenediphenol (8CI), (Chloromethyl)oxirane-4,4'-(1-methylethylidene)bisphenol copolymer, 02Y040, 1-Chloro-2,3-epoxypropane-4,4'-isopropylidenebisphenol polymers, 1-Chloro-2,3-epoxypropane-4,4'-isopropylidenediphenol polymer, 1001X75, 164SV, 2,2-Bis(4-hydroxyphenyl)propane-epichlorohydrin copolymer, 2,2-Bis(4-hydroxyphenyl)propane-epichlorohydrin polymer, 2,2-Bis(hydroxyphenyl)propane-epichlorohydrin copolymer, 2,2-Bis(p-hydroxyphenyl)propane-epichlorohydrin condensate, 2,2-Bis(p-hydroxyphenyl)propane-epichlorohydrin copolymer, 2,2-Bis(p-hydroxyphenyl)propane-epichlorohydrin polymer, 2,2-Diphenylolpropane-epichlorohydrin polymer, 4,4'-Dihydroxydiphenylpropane-epichlorohydrin polymer, 4,4'-Isopropylidenediphenol-1-chloro-2,3-epoxypropane polymer, 4,4'-Isopropylidenediphenol-epichlorohydrin copolymer, 4,4'-Isopropylidenediphenol-epichlorohydrin polymer, Araldite, Bakelite, Biresin, Bis(p-hydroxyphenyl)dimethylmethane-epichlorohydrin copolymer, Bisphenol A epoxy resin, Bisphenol A-(chloromethyl)oxirane copolymer, Bisphenol A-2-(chloromethyl)oxirane copolymer, Bisphenol A-epichlorohydrin condensate, Bisphenol A-epichlorohydrin copolymer, Bisphenol A-epichlorohydrin epoxy resin, Bisphenol A-epichlorohydrin polymer, Bisphenol A-epichlorohydrin resin, Bisphenol A-epichlorohydrin-2-methylimidazole copolymer, Bisphenol a-epichlorohydrin-benzyl alcohol copolymer, Bisphenol a-epichlorohydrin-dibenzylamine copolymer, Bisphenol A-epichlorohydrin-Epikote 1001X75 copolymer, Bisphenol A-epichlorohydrine copolymer, Bisphenol A-Epiclon 850 copolymer, Bisphenol A-Epikote 828 copolymer, Bisphenol A-Epon 829 copolymer, Bisphenol A-glycidyl chloride copolymer, Bisphenol A-p-cumylphenol-epichlorohydrin copolymer, Dian-epichlorohydrin copolymer, Epichlorohydrin-2,2-bis(4-hydroxyphenyl)propane copolymer, Epichlorohydrin-2,2-bis(4-hydroxyphenyl)propane polymer, Epichlorohydrin-4,4'-dihydroxydiphenylpropane copolymer, Epichlorohydrin-4,4'-isopropylidenediphenol copolymer, Epichlorohydrin-4,4'-isopropylidenediphenol polymer, Epichlorohydrin-bisphenol A copolymer, Epichlorohydrin-bisphenol A epoxy resin, Epichlorohydrin-bisphenol A polymer, Epichlorohydrin-bisphenol A resin, Epichlorohydrin-diphenylolpropane polymer, Epiclon, Epidian, Epikote, Epomik, Epon
SMILES CC(C)(c1ccc(O)cc1)c2ccc(O)cc2.ClCC3CO3

Specifications

Quantity 25 g
Formula Weight 320.1179
Chemical Name or Material Epon 828, Technical Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.