missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Adapters for Rotors- 5430/5430 R centrifuge

Product Code. 15178354 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
Large bore hole of 6 x 5mL rotor of Eppendorf Centrifuge 5430/5430 R
Small bore hole of 6 x 5mL rotor of Eppendorf Centrifuge 5430/5430 R
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15178354 Large bore hole of 6 x 5mL rotor of Eppendorf Centrifuge 5430/5430 R Pack of 2
15188354 Small bore hole of 6 x 5mL rotor of Eppendorf Centrifuge 5430/5430 R Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15178354 Supplier Eppendorf™ Supplier No. 5427747001
  • ONLINE EXCLUSIVE PRICE
    105.00 €
    103.00 € / Pack of 2
    Save 2.00 €
    1% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with rotor F-35-6-30

  • For 1 Eppendorf Tube 5.0mL
  • Available rotor bore sizes: large and small

TRUSTED_SUSTAINABILITY

Specifications

Type Adapter
For Use With (Equipment) Large bore hole of 6 x 5mL rotor of Eppendorf Centrifuge 5430/5430 R
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.