Learn More
Eppendorf™ FA-45-24-11-HS Rotor Removal Tool
Shop All Eppendorf Products
Click to view available options
:
Rotor Removal Tool for F-35-6-30 rotor
Description
- Rotor Removal Tool
- For FA-45-24-11-HS rotor with Model 5430/5430R Microcentrifuge
Specifications
Specifications
| Type | Torque Wrench Rotor Key |
| For Use With (Equipment) | Eppendorf, 5430/5430R microcentrifuge with FA-45-24-11-HS 24-place high speed rotor |