missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Rotors for 5430/5430 R Centrifuge
Features aerosol-tight w/ Eppendorf QuickLock™
Brand: Eppendorf™ 5427750002
Code : 99
Additional Details : Weight : 2.25000kg
Description
- Max. speed: 21,191 × g (14,200 rpm)
- Max. capacity: 16 × Eppendorf Tubes™ 5.0 mL
- Aerosol-tight for safe centrifugation of hazardous samples
- Eppendorf QuickLock™ lid
- Eppendorf Tubes™ 5.0 mL with screw caps only can use up half (8) of the places in the rotor
Specifications
Fixed Angle Rotor | |
5430/5430R Centrifuges | |
16 |
17 mm | |
Lid |