missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Top Buret Bottle-Top Burette

Product Code. 10236721 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
50 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10236721 50 mL Each
10236671 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10236721 Supplier Eppendorf™ Supplier No. 4965000017
  • ONLINE EXCLUSIVE PRICE
    1105.50 €
    1000.00 € / Pack of 1
    Save 105.50 €
    9% Off
View all promotions
In Stock
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extremely robust with the display and control elements guaranteeing simple, fatigue-free operation

The innovative Top Buret from Eppendorf sets new standards for manual titration. A new, pulse-free dispensing technique allows the desired volumes to be dispensed continuously, thus eliminating the need for new liquid aspiration each time. This makes titration simpler and safer, with precision values well within required limits

  • Continuous, pulse-free dispensing technique for rapid titration
  • Patented safety valve for deairing without loss of reagent
  • Voltage supply with long-life primary cells and charging display
  • Dispensing range from 0.01mL to 999.9mL
  • Removable electronics unit
  • Single channel with simple calibration program
  • Discharge tube rotatable by 360°
  • Adjustable discharge tube: horizontal: 142 to 220mm, vertical: 10 to 200mm

  • Telescopic aspiration tube for automatically setting the length from 210mm to 370mm
  • Connection for dry tube
  • The medium is dispensed by simply rotating the wheels

Model M

  • Dispensing rate of 2,500μL per rotation
Model H
  • Dispensing rate of 5,000μL per rotation

TRUSTED_SUSTAINABILITY

Specifications

Accuracy Standard
Product Type Digital Bottletop Dispenser
Volume (Metric) 50 mL
For Use With (Equipment) Rapid Titration
Includes Recirculation valve, valve toggle, telescopic filling tube, adjustable discharge tube, three adapters 40/38/32 mm bottle threads, 2 x 1.5 V micro-battery, quality certificate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.