missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Vis Cuvettes
Disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm. The Vis cuvettes are the perfect tool for applications outside of the UV range, for example, colorimetric protein assays (Bradford, Lowry, etc.), determining of the optical density of bacterial cultures (OD 600 methods), kinetic, and fluorescence measurements.
149.00 € - 162.02 €
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15234566
|
Eppendorf™
0030079353 |
Semi-micro Vis Cuvet |
162.02 €
Pack of 1000 |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15273993
|
Eppendorf™
0030079345 |
Macro Vis Cuvet |
149.00 €
Pack of 1000 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Two different cuvette sizes (semi-micro and macro) for a wide variety of volumes and applications
- Disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm
Specifications
Plastic |