missing translation for 'onlineSavingsMsg'
Learn More

EPPS, 99%

Product Code. 11494714
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494714 100 g 100g
11484714 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494714 Supplier Thermo Scientific Alfa Aesar Supplier No. A13714.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16052-06-5 | C9H20N2O4S | 252.329 g/mol

EPPS is used as a biological buffer with a useful pH range of 7.3-8.7 & pKa=8.0 (25°C). It can be used in agrochemical, pharmaceutical and dyestuff field etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
EPPS is used as a biological buffer with a useful pH range of 7.3-8.7 & pKa=8.0 (25°C). It can be used in agrochemical, pharmaceutical and dyestuff field etc.

Solubility
Soluble in water. 25.2 g/L (20°C)

Notes
Store at room temperature. Incompatible with oxidizing agents. Store away from extreme heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16052-06-5
Molecular Formula C9H20N2O4S
Molecular Weight (g/mol) 252.329
MDL Number MFCD00006160
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
Synonym hepps, epps, 4-2-hydroxyethyl-1-piperazinepropanesulfonic acid, 3-4-2-hydroxyethyl piperazin-1-yl propane-1-sulfonic acid, 1-piperazinepropanesulfonic acid, 4-2-hydroxyethyl, n-2-hydroxyethyl piperazine-n'-3-propanesulfonic acid, 4-2-hydroxyethyl piperazine-1-propanesulfonic acid, 4-2-hydroxyethyl-1-piperazinepropane sulfonic acid, 4-2-hydroxyethyl-1-piperazinepropanesulphonic acid
PubChem CID 85255
ChEBI CHEBI:42298
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO

Specifications

Melting Point 242°C (decomposition)
Quantity 100 g
Beilstein 3957385
Solubility Information Soluble in water. 25.2g/L (20°C)
Formula Weight 252.33
Percent Purity 99%
Chemical Name or Material EPPS

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.