missing translation for 'onlineSavingsMsg'
Learn More

Eprinomectin, MedChemExpress

Product Code. 16475936
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16475936 100 mg 100mg
16431646 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16475936 Supplier Medchem Express Supplier No. HY12643
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Eprinomectin(MK-397) is an avermectin selected for development as a topical endectocide; has anthelmintic, insecticidal and miticidal activity.

Eprinomectin(MK-397) is an avermectin selected for development as a topical endectocide

Specifications

Chemical Name or Material Eprinomectin
CAS 123997-26-2
Color White
Purity Grade Notes Research
Molecular Formula C50H75NO14
Quantity 100 mg
Synonym MK-397
Solubility Information DMSO : ≥ 100 mg/mL (109.39 mM)
SMILES C[C@H]1[C@@H](NC(C)=O)[C@@H](OC)C[C@H](O[C@@H]2[C@H](C)O[C@@H](O[C@@H](/C(C)=C/C[C@@H](C3)OC4(O[C@@H]([C@@H](CC)C)[C@H](C)C=C4)C[C@H]3OC5=O)[C@@H](/C=C/C=C6CO[C@@H]7[C@@]\6(O)[C@H]5C=C(C)[C@H]7O)C)C[C@@H]2OC)O1
Molecular Weight (g/mol) 914.13
Formula Weight 914.13
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.