missing translation for 'onlineSavingsMsg'
Learn More

Epristeride, MedChemExpress

Product Code. 16496817
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16496817

Brand: Medchem Express HY107385

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Epristeride is a novel 5α-reductase inhibor.

Epristeride is a novel 5α-reductase inhibor.

Specifications

Chemical Name or Material Epristeride
CAS 119169-78-7
Color White
Purity Grade Notes Research
Molecular Formula C25H37NO3
Quantity 5 mg
Synonym ONO-9302 SKF105657
Solubility Information DMSO : 100 mg/mL (250.27 mM; Need ultrasonic)
SMILES C[C@@]12[C@](CC[C@@H]2C(NC(C)(C)C)=O)([H])[C@@]3([H])[C@]([C@@]4(C(C=C(C(O)=O)CC4)=CC3)C)([H])CC1
Molecular Weight (g/mol) 399.57
Formula Weight 399.57
Percent Purity 98.39%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.