missing translation for 'onlineSavingsMsg'
Learn More

epsilon-Caprolactam, TRC

Product Code. 30134690
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Unit Size:
250g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134690 250 g 250g
30003274 5 g 5g
30004598 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134690 Supplier Toronto Research Chemicals Supplier No. TRCC175665250G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-60-2
Molecular Formula C6 H11 N O
Molecular Weight (g/mol) 113.16
Synonym 2H-Azepin-2-one, hexahydro-, Hexahydro-2H-azepin-2-one, 1-Aza-2-cycloheptanone, 2-Azacycloheptanone, 2-Ketohexamethylenimine, 2-Oxoazepane, 2-Oxohexamethylenimine, 2-Perhydroazepinone, 6-Caprolactam, 6-Hexanelactam, A 19374, AP, AP (lactam), Aminocaproic lactam, Azepan-2-one, Caprolactam, Hexahydro-1H-azepin-2-one, Hexahydro-2-azepinone, Hexano-6-lactam, Hexanoic acid, 6-amino-, cyclic lactam, Hexanolactam, NSC 117393, NSC 25536, NSC 4977, ε-Caprolactam, ω-Caprolactam
IUPAC Name azepan-2-one
SMILES O=C1CCCCCN1

Specifications

Quantity 250 g
InChI Formula InChI=1S/C6H11NO/c8-6-4-2-1-3-5-7-6/h1-5H2,(H,7,8)
Formula Weight 113.0841
Percent Purity >95%
Chemical Name or Material e-Caprolactam
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.