missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ Epstein-Barr Virus Antigen Microarray

Product Code. 16317758
Change view
Click to view available options
Quantity: 1 Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16317758 Quantity: 1 Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16317758 Supplier PEPperCHIP™ Supplier No. SPC.008.001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Epstein-Barr Virus Antigen Microarray with 4101 virus specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.

  • Microarray Content: 13 selected antigens of Epstein-Barr virus strain B95-8: Gp350, EBNA1, EBNA2, EBNA3A, EBNA3B, EBNA3C, EBNA-LP, LMP1, LMP2, BZLF1 (Zebra), BRLF1 (Rta), BMLF1, and BMRF1
  • Peptide Length/Overlap: 15 aa/ 13 aa
  • Number of Peptides: 4101 printed in duplicate and HA, polio and c-Myc control peptides

Specifications

Coating PEGMA/PMMA
Description 1 array copy per microarray
Quantity 1 Slide
Method of Attachment Combinatorial peptide synthesis by amino acid laser printing
Thickness (Metric) 1 mm
Certifications/Compliance ISO 9001:2015
Color Transparent
Regulatory Status RUO
Barcode Individual barcode for each microarray
Material Microscopic Glass
Type High-density Peptide Microarray
For Use With (Application) Antibody validation, epitope mapping, serum screening, biomarker discovery
Sterility Non-sterile
Dimensions (LxW) 75.4 x 25 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.